1,2,3,5-Tetramethylbenzene ≥70.0%

Supplier: TCI America

Synonyms: Isodurene

T0565-25ML T0565-5ML
TCT0565-5MLEA 151.48 USD
TCT0565-5ML TCT0565-025ML
1,2,3,5-Tetramethylbenzene ≥70.0%
1,2,3,5-Tetramethylbenzene

CAS Number: 527-53-7
MDL Number: MFCD00008524
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: 70.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 198
Flash Point (°C): 63
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Isodurene

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 198 °C
Melting Pt: –23.7 °C
Density: 0.89 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00008524
CAS Number: 527-53-7
EINECS: 208-417-1
Merck Index: 12,05182
https://us.vwr.com/stibo/web/std.lang.all/47/84/28994784.jpg

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR