Triphenylantimony ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Triphenylstibine

T0476-25G
TCT0476-025GEA 65.96 USD
TCT0476-025G
Triphenylantimony ≥95.0% (by titrimetric analysis)
Triphenylantimony

CAS Number: 603-36-1
MDL Number: MFCD00003000
Molecular Formula: C18H15Sb
Molecular Weight: 353.08
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 377
Melting point (°C): 53
Flash Point (°C): 110


Other Product Names / Synonyms: Triphenylstibine

Formula: C₁₈H₁₅Sb
MW: 353.08 g/mol
Boiling Pt: 377 °C
Melting Pt: 53 °C
Density: 1.5 g/cm³ (12 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003000
CAS Number: 603-36-1
EINECS: 210-037-6
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/47/30/28994730.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR