TCIAT0359

T0359-25G
TCT0359-25GEA 27.23 USD
TCT0359-25G
TCIAT0359
CAS Number: 732-26-3
MDL Number: MFCD00008821
Molecular Formula: C18H30O
Molecular Weight: 262.44
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 131
Flash Point (°C): 130
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR