Methyl thiocyanate ≥99.0%

Supplier: TCI America

T0201-25ML T0201-500ML
TCT0201-500MLEA 402.83 USD
TCT0201-500ML TCT0201-025ML
Methyl thiocyanate ≥99.0%
Methyl thiocyanate

CAS Number: 556-64-9
MDL Number: MFCD00001830
Molecular Formula: C2H3NS
Molecular Weight: 73.11
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 130
Flash Point (°C): 38
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: Methyl Rhodanide, Thiocyanic Acid Methyl Ester

Formula: C₂H₃NS
MW: 73.12 g/mol
Boiling Pt: 130 °C
Melting Pt: –5 °C
Density: 1.08 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00001830
CAS Number: 556-64-9
EINECS: 209-134-6
UN: 2929
ADR: 6.1,II
Merck Index: 12,06209
https://us.vwr.com/stibo/web/std.lang.all/45/18/28994518.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR