Myristoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Tetradecanoyl chloride

T0086-25G
TCT0086-025GEA 50.22 USD
TCT0086-025G
Myristoyl chloride ≥98.0% (by GC, titration analysis)
Myristoyl chloride

CAS Number: 112-64-1
MDL Number: MFCD00000741
Molecular Formula: C14H27ClO
Molecular Weight: 246.82
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 165
Flash Point (°C): 110
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: Tetradecanoyl Chloride

Formula: C₁₄H₂₇ClO
MW: 246.82 g/mol
Boiling Pt: 165 °C
Density: 0.91 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000741
CAS Number: 112-64-1
EINECS: 203-994-6
UN: 3265
ADR: 8,I
https://us.vwr.com/stibo/web/std.lang.all/44/19/28994419.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR