Strychnine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

S0249-25G
TCS0249-025GEA 65 USD
TCS0249-025G
Strychnine ≥98.0% (by HPLC, titration analysis)
Strychnine

CAS Number: 57-24-9
MDL Number: MFCD00005941
Molecular Formula: C21H22N2O2
Molecular Weight: 334.42
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White

Formula: C₂₁H₂₂N₂O₂
MW: 334.42 g/mol
MDL Number: MFCD00005941
CAS Number: 57-24-9
EINECS: 200-319-7
https://us.vwr.com/stibo/web/std.lang.all/40/04/28994004.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR