TCIAS0235

S0235-500G
TCS0235-500GEA 49.96 USD
TCS0235-500G
TCIAS0235
CAS Number: 6865-35-6
MDL Number: MFCD00050711
Molecular Formula: C18H36O2
Molecular Weight: 704.28
Form: Crystal
Color: White
Melting point (°C): 120

Other Product Names / Synonyms: Stearic Acid Barium Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR