7-Quinolinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 7-HydroxyquinolineQuinolin-7-ol

Q0084-1G
TCQ0084-1GEA 180 USD
TCQ0084-1G
7-Quinolinol ≥98.0% (by GC, titration analysis)
7-Quinolinol

CAS Number: 580-20-1
MDL Number: MFCD00016730
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 242


Other Product Names / Synonyms: 7-Hydroxyquinoline

Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 242 °C
MDL Number: MFCD00016730
CAS Number: 580-20-1
EINECS: 209-457-2
https://us.vwr.com/stibo/web/std.lang.all/37/21/28993721.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR