1-Phenylisoquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1731-5G P1731-1G
TCP1731-5GEA 430.87 USD
TCP1731-5G TCP1731-1G
1-Phenylisoquinoline ≥98.0% (by GC, titration analysis)
1-Phenylisoquinoline

CAS Number: 3297-72-1
MDL Number: MFCD00093934
Molecular Formula: C15H11N
Molecular Weight: 205.26
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 130
Melting point (°C): 96

Formula: C₁₅H₁₁N
Boiling Pt: 130 °C
Melting Pt: 96 °C
CAS Number: 3297-72-1
https://us.vwr.com/stibo/web/std.lang.all/30/54/28993054.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR