Phenyl chlorodithioformate ≥97.0%

Supplier: TCI America

Synonyms: Chlorodithioformic acid Phenyl ester

P1266-5G
TCP1266-005GEA 102 USD
TCP1266-005G
Phenyl chlorodithioformate ≥97.0%
Phenyl chlorodithioformate

CAS Number: 16911-89-0
MDL Number: MFCD00013429
Molecular Formula: C7H5ClS2
Molecular Weight: 188.69
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Yellow
Boiling point (°C): 95
Flash Point (°C): 113
Specific Gravity (20/20): 1.32
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Chlorodithioformic Acid Phenyl Ester

Formula: C₇H₅ClS₂
MW: 188.7 g/mol
Boiling Pt: 95 °C
Density: 1.32 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00013429
CAS Number: 16911-89-0
https://us.vwr.com/stibo/web/std.lang.all/18/97/28991897.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR