9-Phenylacridine ≥98.0%

Supplier: TCI America

P1198-10G
TCP1198-010GEA 132 USD
TCP1198-010G
9-Phenylacridine ≥98.0%
9-Phenylacridine

CAS Number: 602-56-2
MDL Number: MFCD00092562
Molecular Formula: C19H13N
Molecular Weight: 255.32
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 184

Formula: C₁₉H₁₃N
MW: 255.32 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00092562
CAS Number: 602-56-2
EINECS: 210-020-3
https://us.vwr.com/stibo/web/std.lang.all/18/33/28991833.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR