Diethyl pimelate ≥97.0%
Supplier: TCI America
Synonyms:
Pimelic acid diethyl ester
, Heptanedioic acid diethyl ester
, Diethyl Heptanedioate
CAS Number: 2050-20-6
MDL Number: MFCD00009216
Molecular Formula: C11H20O4
Molecular Weight: 216.28
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 149
Flash Point (°C): 110
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Diethyl Heptanedioate, Pimelic Acid Diethyl Ester, Heptanedioic Acid Diethyl Ester
Formula:
C₁₁H₂₀O₄ MW: 216.28 g/mol Boiling Pt: 149 °C Melting Pt: –24 °C Density: 0.99 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009216 CAS Number: 2050-20-6 EINECS: 218-083-9 Merck Index: 12,07585 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...