2-Piperidinemethanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(Hydroxymethyl)piperidine

P1017-25G
TCP1017-025GEA 57.2 USD
TCP1017-025G
2-Piperidinemethanol ≥98.0% (by GC, titration analysis)
2-Piperidinemethanol

CAS Number: 3433-37-2
MDL Number: MFCD00005987
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 100
Melting point (°C): 67


Other Product Names / Synonyms: 2-(Hydroxymethyl)piperidine

Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 100 °C
Melting Pt: 67 °C
MDL Number: MFCD00005987
CAS Number: 3433-37-2
EINECS: 222-333-2
https://us.vwr.com/stibo/web/std.lang.all/16/54/28991654.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR