Pentacosanoic acid ≥96.0%

Supplier: TCI America

P0882-5G P0882-1G
TCP0882-005GEA 692 USD
TCP0882-005G TCP0882-001G
Pentacosanoic acid ≥96.0%
Pentacosanoic acid

CAS Number: 506-38-7
MDL Number: MFCD00020551
Molecular Formula: C25H50O2
Molecular Weight: 382.67
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 83

Formula: C₂₅H₅₀O₂
MW: 382.67 g/mol
Melting Pt: 83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00020551
CAS Number: 506-38-7
EINECS: 208-036-0
https://us.vwr.com/stibo/web/std.lang.all/15/51/28991551.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR