1,2-Propanedithiol ≥95.0%

Supplier: TCI America

Synonyms: Propylenedithiol1,2-Dimercaptopropane

P0821-25ML P0821-5ML
TCP0821-005MLEA 249 USD
TCP0821-005ML TCP0821-025ML
1,2-Propanedithiol ≥95.0%
1,2-Propanedithiol

CAS Number: 814-67-5
MDL Number: MFCD00022073
Molecular Formula: C3H8S2
Molecular Weight: 108.22
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 35


Other Product Names / Synonyms: 1,2-Dimercaptopropane, Propylenedithiol

Formula: C₃H₈S₂
MW: 108.23 g/mol
Flash Pt: 35 °C
MDL Number: MFCD00022073
CAS Number: 814-67-5
EINECS: 212-398-5
https://us.vwr.com/stibo/web/std.lang.all/14/91/28991491.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR