4-Pentyn-2-ol ≥97.0%

Supplier: TCI America
Danger

Synonyms: 1-Pentyn-4-ol

P0818-5ML
TCP0818-005MLEA 55.83 USD
TCP0818-005ML
4-Pentyn-2-ol ≥97.0%
4-Pentyn-2-ol

CAS Number: 2117-11-5
MDL Number: MFCD00004555
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 37
Specific Gravity (20/20): 0.90

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 126 °C
Density: 0.90 (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00004555
CAS Number: 2117-11-5
EINECS: 218-321-1
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR