Pentadecylbenzene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Phenylpentadecane

P0665-25ML
TCP0665-025MLEA 525 USD
TCP0665-025ML
Pentadecylbenzene ≥98.0%
Pentadecylbenzene

CAS Number: 2131-18-2
MDL Number: MFCD00015083
Molecular Formula: C21H36
Molecular Weight: 288.52
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 373
Freezing point (°C): 12
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 1-Phenylpentadecane

Formula: C₂₁H₃₆
MW: 288.52 g/mol
Boiling Pt: 373 °C
Melting Pt: 22 °C
Density: 0.86 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015083
CAS Number: 2131-18-2
EINECS: 218-352-0
https://us.vwr.com/stibo/web/std.lang.all/13/78/28991378.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR