Propionyl chloride ≥98.0% (by GC, titration analysis)

Danger

P0516-500G P0516-25G
TCP0516-500GEA 45.9 USD
TCP0516-500G TCP0516-025G
Propionyl chloride ≥98.0% (by GC, titration analysis)
Propionyl chloride

CAS Number: 79-03-8
MDL Number: MFCD00000745
Molecular Formula: C3H5ClO
Molecular Weight: 92.52
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 80
Melting point (°C): -94
Flash Point (°C): 12
Specific Gravity (20/20): 1.06

Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 80 °C
Melting Pt: –94 °C
Density: 1.06 (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00000745
CAS Number: 79-03-8
EINECS: 201-170-0
UN: 1815
ADR: 3,II
Merck Index: 12,08013
https://us.vwr.com/stibo/web/std.lang.all/12/27/28991227.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR