2-Propylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-n-Propylpyridine

P0336-25ML
TCP0336-025MLEA 198 USD
TCP0336-025ML
2-Propylpyridine ≥98.0% (by GC, titration analysis)
2-Propylpyridine

CAS Number: 622-39-9
MDL Number: MFCD00006368
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 167
Flash Point (°C): 53
Specific Gravity (20/20): 0.92

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 167 °C
Melting Pt: 1 °C
Density: 0.92 (20 °C)
Flash Pt: 53 °C
MDL Number: MFCD00006368
CAS Number: 622-39-9
EINECS: 210-732-4
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/10/66/28991066.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR