N-Phenyl-2-naphthylamine ≥97.0% (by total nitrogen basis)

Supplier: TCI America

P0198-25G P0198-500G
TCP0198-500GEA 176 USD
TCP0198-500G TCP0198-025G
N-Phenyl-2-naphthylamine ≥97.0% (by total nitrogen basis)
N-Phenyl-2-naphthylamine

CAS Number: 135-88-6
MDL Number: MFCD00004052
Molecular Formula: C16H13N
Molecular Weight: 219.29
Purity/Analysis Method: 97.0% (N)
Form: Crystal
Boiling point (°C): 237
Melting point (°C): 108


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 2-Anilinonaphthalene

Formula: C₁₆H₁₃N
Boiling Pt: 237 °C
Melting Pt: 108 °C
MDL Number: MFCD00004052
CAS Number: 135-88-6
EINECS: 205-223-9
https://us.vwr.com/stibo/web/std.lang.all/09/31/28990931.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR