4-Aminoazobenzene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-(Phenylazo)aniline
P0141-25G
P0141-100G
TCP0141-025GEA
107.23
USD
TCP0141-025G
TCP0141-100G
4-Aminoazobenzene ≥98.0% (by HPLC, titration analysis)
4-Aminoazobenzene
CAS Number: 60-09-3
MDL Number: MFCD00007854
Molecular Formula: C12H11N3
Molecular Weight: 197.24
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 125
Other Product Names / Synonyms: 4-(Phenylazo)aniline
Formula:
C₁₂H₁₁N₃ MW: 197.24 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00007854 CAS Number: 60-09-3 EINECS: 200-453-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...