2-Oxobutyric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2-Ketobutyric acidα-Ketobutyric acid2-Oxobutanoic acid

O0364-5G
TCO0364-5GEA 423 USD
TCO0364-5G
2-Oxobutyric acid ≥98.0% (by GC, titration analysis)
2-Oxobutyric acid

CAS Number: 600-18-0
MDL Number: MFCD00004164
Molecular Formula: C4H6O3
Molecular Weight: 102.09
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 84
Melting point (°C): 32
Flash Point (°C): 82
Storage Temperature: 0-10°C

Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 84 °C
Melting Pt: 32 °C
Density: 1.2 g/cm³ (17 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004164
CAS Number: 600-18-0
EINECS: 209-986-9
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/05/95/28990595.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR