cis-3-Octene ≥98.0%

Supplier: TCI America

Synonyms: (Z)-3-Octene

O0133-5ML
TCO0133-005MLEA 633.09 USD
TCO0133-005ML
cis-3-Octene ≥98.0%
cis-3-Octene

CAS Number: 14850-22-7
MDL Number: MFCD00067486
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 17
Specific Gravity (20/20): 0.72

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 123 °C
Melting Pt: –126 °C
Density: 0.72 (20 °C)
Flash Pt: 17 °C
MDL Number: MFCD00067486
CAS Number: 14850-22-7
EINECS: 238-912-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR