1-Octadecyne ≥95.0%
Supplier: TCI America
O0128-5ML
O0128-1ML
76220-388EA
57.86
USD
76220-388
TCO0128-005ML
1-Octadecyne ≥95.0%
1-Octadecyne
CAS Number: 629-89-0
MDL Number: MFCD00015088
Molecular Formula: C18H34
Molecular Weight: 250.47
Purity/Analysis Method:
Form: Crystal
Freezing point (°C): 25
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Cetylacetylene, Hexadecylacetylene
Formula:
C₁₈H₃₄ MW: 250.47 g/mol Boiling Pt: 315 °C (1013 hPa) Melting Pt: 28 °C Density: 0.8696 g/cm³ (0 °C) |
MDL Number:
MFCD00015088 CAS Number: 629-89-0 EINECS: 211-114-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...