Nonacosanoic acid ≥95.0%

Supplier: TCI America

N0662-5G N0662-1G
TCN0662-5GEA 449.45 USD
TCN0662-5G TCN0662-1G
Nonacosanoic acid ≥95.0%
Nonacosanoic acid

CAS Number: 4250-38-8
MDL Number: MFCD00057830
Molecular Formula: C29H58O2
Molecular Weight: 438.78
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 90

Formula: C₂₉H₅₈O₂
MW: 438.78 g/mol
Melting Pt: 90 °C
MDL Number: MFCD00057830
CAS Number: 4250-38-8
EINECS: 224-210-9
https://us.vwr.com/stibo/web/std.lang.all/98/81/28989881.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR