trans,trans-2,4-Nonadienal ≥85.0%
Supplier: TCI America
N0521-5ML
N0521-25ML
TCN0521-25MLEA
175.44
USD
TCN0521-25ML
TCN0521-005ML
trans,trans-2,4-Nonadienal ≥85.0%
trans,trans-2,4-Nonadienal
CAS Number: 5910-87-2
MDL Number: MFCD00007006
Molecular Formula: C9H14O
Molecular Weight: 138.21
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 90
Specific Gravity (20/20): 0.87
Formula:
C₉H₁₄O MW: 138.21 g/mol Boiling Pt: 99 °C Density: 0.87 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00007006 CAS Number: 5910-87-2 EINECS: 227-629-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...