Nonadecylcyclohexane ≥98.0%
Supplier: TCI America
Synonyms:
1-Cyclohexylnonadecane
N0394-25G
N0394-5G
TCN0394-025GEA
685
USD
TCN0394-025G
TCN0394-5G
Nonadecylcyclohexane ≥98.0%
Nonadecylcyclohexane
CAS Number: 22349-03-7
MDL Number: MFCD00058954
Molecular Formula: C25H50
Molecular Weight: 350.68
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 420
Melting point (°C): 45
Other Product Names / Synonyms: 1-Cyclohexylnonadecane
Formula:
C₂₅H₅₀ MW: 350.67 g/mol Boiling Pt: 420 °C Melting Pt: 45 °C Density: 0.831 g/cm³ (20 °C) |
MDL Number:
MFCD00058954 CAS Number: 22349-03-7 EINECS: 244-930-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...