Nitrosobenzene ≥98.0%

Supplier: TCI America

N0367-25G N0367-5G
TCN0367-025GEA 225 USD
TCN0367-025G TCN0367-5G
Nitrosobenzene ≥98.0%
Nitrosobenzene

CAS Number: 586-96-9
MDL Number: MFCD00002059
Molecular Formula: C6H5NO
Molecular Weight: 107.11
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 57
Melting point (°C): 66
Storage Temperature: 0-10°C

Formula: C₆H₅NO
MW: 107.11 g/mol
Boiling Pt: 57 °C
Melting Pt: 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002059
CAS Number: 586-96-9
EINECS: 209-591-1
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/94/50/28989450.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR