3-Nonyne ≥98.0%

Supplier: TCI America

Synonyms: EthylpentylacetyleneAmylethylacetylene

N0319-5ML
TCN0319-005MLEA 170.44 USD
TCN0319-005ML
3-Nonyne ≥98.0%
3-Nonyne

CAS Number: 20184-89-8
MDL Number: MFCD00039977
Molecular Formula: C9H16
Molecular Weight: 124.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 156
Specific Gravity (20/20): 0.76


Other Product Names / Synonyms: Amylethylacetylene, Ethylpentylacetylene

Formula: C₉H₁₆
MW: 124.23 g/mol
Boiling Pt: 156 °C
Density: 0.76 (20 °C)
MDL Number: MFCD00039977
CAS Number: 20184-89-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR