1-Nonene ≥90.0%

N0295-25ML
TCN0295-025MLEA 84.36 USD
TCN0295-025ML
1-Nonene ≥90.0%
1-Nonene

CAS Number: 124-11-8
MDL Number: MFCD00009568
Molecular Formula: C9H18
Molecular Weight: 126.24
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 26
Specific Gravity (20/20): 0.73

Formula: C₉H₁₈
MW: 126.24 g/mol
Boiling Pt: 147 °C
Melting Pt: –88 °C
Density: 0.73 (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009568
CAS Number: 124-11-8
EINECS: 204-681-7
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR