2-Nitrofluorene ≥99.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-Nitro-9H-fluorene

N0201-100G N0201-25G
TCN0201-025GEA 86.28 USD
TCN0201-025G TCN0201-100G
2-Nitrofluorene ≥99.0% (by HPLC)
2-Nitrofluorene

CAS Number: 607-57-8
MDL Number: MFCD00001117
Molecular Formula: C13H9NO2
Molecular Weight: 211.22
Purity/Analysis Method: 99.0% (HPLC)
Form: Crystal
Melting point (°C): 155

Formula: C₁₃H₉NO₂
MW: 211.22 g/mol
Melting Pt: 155 °C
MDL Number: MFCD00001117
CAS Number: 607-57-8
EINECS: 210-138-5
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/92/90/28989290.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR