Nitrocyclohexane ≥95.0%
Supplier: TCI America
N0187-5G
N0187-25G
TCN0187-005GEA
115.57
USD
TCN0187-005G
TCN0187-025G
Nitrocyclohexane ≥95.0%
Nitrocyclohexane
CAS Number: 1122-60-7
MDL Number: MFCD00003843
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 113
Melting point (°C): -34
Flash Point (°C): 74
Specific Gravity (20/20): 1.06
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Boiling Pt: 113 °C Melting Pt: –34 °C Density: 1.06 (20 °C) Flash Pt: 74 °C |
MDL Number:
MFCD00003843 CAS Number: 1122-60-7 EINECS: 214-354-0 UN: 3382 ADR: 6.1,I |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...