2-Nitrophenylarsonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: 2-Nitrobenzenearsonic acid

N0133-25G N0133-1G
TCN0133-025GEA 158.26 USD
TCN0133-025G TCN0133-001G
2-Nitrophenylarsonic acid ≥98.0% (by titrimetric analysis)
2-Nitrophenylarsonic acid

CAS Number: 5410-29-7
MDL Number: MFCD00047664
Molecular Formula: C6H6AsNO5
Molecular Weight: 247.04
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Nitrobenzenearsonic Acid

Formula: C₆H₆AsNO₅
MW: 247.04 g/mol
MDL Number: MFCD00047664
CAS Number: 5410-29-7
EINECS: 226-485-0
UN: 3465
ADR: 6.1,I
https://us.vwr.com/stibo/web/std.lang.all/92/30/28989230.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR