3-Nitroanisole ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1-Methoxy-3-nitrobenzenem-Nitroanisole

N0124-25G
TCN0124-025GEA 192.79 USD
TCN0124-025G
3-Nitroanisole ≥99.0%
3-Nitroanisole

CAS Number: 555-03-3
MDL Number: MFCD00007237
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 36
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Methoxy-3-nitrobenzene

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 32…35 °C
Density: 1.37 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007237
CAS Number: 555-03-3
EINECS: 209-079-8
UN: 3458
ADR: 6.1,III
Merck Index: 12,06682
https://us.vwr.com/stibo/web/std.lang.all/92/23/28989223.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR