9-Methylacridine ≥97.0%

Supplier: TCI America

M1237-1G M1237-5G
TCM1237-5GEA 362.25 USD
TCM1237-5G TCM1237-001G
9-Methylacridine ≥97.0%
9-Methylacridine

CAS Number: 611-64-3
MDL Number: MFCD00143523
Molecular Formula: C14H11N
Molecular Weight: 193.25
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 117

Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00143523
CAS Number: 611-64-3
EINECS: 210-272-4
https://us.vwr.com/stibo/web/std.lang.all/50/25/28975025.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR