7-Methylcoumarin ≥98.0%

Supplier: TCI America

Synonyms: 7-Methyl-2-benzopyrone

M1236-25G
TCM1236-025GEA 228 USD
TCM1236-025G
7-Methylcoumarin ≥98.0%
7-Methylcoumarin

CAS Number: 2445-83-2
MDL Number: MFCD00006880
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 128


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 7-Methylcumarin

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 128 °C
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00006880
CAS Number: 2445-83-2
EINECS: 219-499-3
https://us.vwr.com/stibo/web/std.lang.all/50/23/28975023.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR