1-Methylpyrene ≥94.0%

Supplier: TCI America
Warning

M0620-100MG
TCM0620-100MGEA 155.23 USD
TCM0620-100MG
1-Methylpyrene ≥94.0%
1-Methylpyrene

CAS Number: 2381-21-7
MDL Number: MFCD00014329
Molecular Formula: C17H12
Molecular Weight: 216.28
Purity/Analysis Method: 94.0% (GC)
Form: Crystal
Boiling point (°C): 372
Melting point (°C): 70

Formula: C₁₇H₁₂
MW: 216.28 g/mol
Boiling Pt: 372 °C
Melting Pt: 70 °C
MDL Number: MFCD00014329
CAS Number: 2381-21-7
EINECS: 219-178-8
https://us.vwr.com/stibo/web/std.lang.all/40/82/28974082.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR