Methyl green (Basic Blue 20)

Supplier: TCI America

Synonyms: Basic Blue 20Double greenMethyl green chlorideLight green

M0498-25G
TCM0498-025GEA 217.15 USD
TCM0498-025G
Methyl green (Basic Blue 20)
Methyl Green

CAS Number: 82-94-0
MDL Number: MFCD00011879
Molecular Formula: C26H33Cl2N3
Molecular Weight: 458.47
Form: Crystal
Color: Deep Green


Other Product Names / Synonyms: Basic Blue 20

Formula: C₂₆H₃₃Cl₂N₃
MW: 458.47 g/mol
MDL Number: MFCD00011879
CAS Number: 82-94-0
EINECS: 201-447-6
https://us.vwr.com/stibo/web/std.lang.all/39/61/28973961.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR