trans-2-Methylcyclohexanol ≥98.0%

Supplier: TCI America
Warning

M0352-5ML
TCM0352-005MLEA 390.78 USD
TCM0352-005ML
trans-2-Methylcyclohexanol ≥98.0%
trans-2-Methylcyclohexanol

CAS Number: 7443-52-9
MDL Number: MFCD00064168
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Melting point (°C): -21
Flash Point (°C): 59
Specific Gravity (20/20): 0.93

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 167 °C
Melting Pt: –21 °C
Density: 0.93 (20 °C)
Flash Pt: 59 °C
MDL Number: MFCD00064168
CAS Number: 7443-52-9
EINECS: 231-186-3
UN: 2617
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR