4-Methyl-2-pentyne ≥96.0%

Supplier: TCI America

M0270-5ML
TCM0270-005MLEA 197 USD
TCM0270-005ML
4-Methyl-2-pentyne ≥96.0%
4-Methyl-2-pentyne

CAS Number: 21020-27-9
MDL Number: MFCD00041615
Molecular Formula: C6H10
Molecular Weight: 82.15
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 72
Flash Point (°C): -4
Specific Gravity (20/20): 0.72


Other Product Names / Synonyms: Isopropylmethylacetylene

Formula: C₆H₁₀
Boiling Pt: 72 °C
Density: 0.72 (20 °C)
Flash Pt: –4 °C
MDL Number: MFCD00041615
CAS Number: 21020-27-9

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR