7-Methylbenz[a]anthracene ≥97.0%

Supplier: TCI America
Warning

M0248-100MG
TCM0248-100MGEA 204 USD
TCM0248-100MG
7-Methylbenz[a]anthracene ≥97.0%
7-Methylbenz[a]anthracene

CAS Number: 2541-69-7
MDL Number: MFCD00059500
Molecular Formula: C19H14
Molecular Weight: 242.32
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 140

Formula: C₁₉H₁₄
MW: 242.32 g/mol
Boiling Pt: 411 °C (1013 hPa)
Melting Pt: 140 °C
MDL Number: MFCD00059500
CAS Number: 2541-69-7
EINECS: 219-819-1
https://us.vwr.com/stibo/web/std.lang.all/36/62/28973662.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR