Methoxyacetonitrile ≥98.0%
Supplier: TCI America
M0103-5ML
M0103-25ML
TCM0103-005MLEA
66.99
USD
TCM0103-005ML
TCM0103-025ML
Methoxyacetonitrile ≥98.0%
Methoxyacetonitrile
CAS Number: 1738-36-9
MDL Number: MFCD00001892
Molecular Formula: C3H5NO
Molecular Weight: 71.08
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 32
Specific Gravity (20/20): 0.95
Formula:
C₃H₅NO Boiling Pt: 119 °C Density: 0.95 (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD00001892 CAS Number: 1738-36-9 EINECS: 217-092-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...