TCIAI0448

I0448-25ML
TCI0448-025MLEA 76.66 USD
TCI0448-025ML
TCIAI0448
CAS Number: 41406-00-2
MDL Number: MFCD00052700
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 245
Specific Gravity (20/20): 1.03
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR