Propyl isobutyrate ≥97.0%

Supplier: TCI America

Synonyms: Isobutyric acid Propyl ester

I0108-25ML
TCI0108-025MLEA 37.98 USD
TCI0108-025ML
Propyl isobutyrate ≥97.0%
Propyl isobutyrate

CAS Number: 644-49-5
MDL Number: MFCD00053781
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 30
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Isobutyric Acid Propyl Ester

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 134 °C
Density: 0.86 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00053781
CAS Number: 644-49-5
EINECS: 211-417-4
https://us.vwr.com/stibo/web/std.lang.all/18/92/28971892.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR