TCIAH1070

H1070-100MG
TCH1070-100MGEA 464.22 USD
TCH1070-100MG
TCIAH1070
CAS Number: 296-35-5
MDL Number: MFCD00042750
Molecular Formula: C12H30N6
Molecular Weight: 258.41
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 149
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR