2,2,4,4,6,6-Hexamethylcyclotrisilazane ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Dimethylsilazane Cyclic trimer2,2,4,4,6,6-Hexamethyl-1,3,5,2,4,6-triazatrisilinane

H0909-5ML H0909-25ML
TCH0909-025MLEA 78.98 USD
TCH0909-025ML TCH0909-005ML
2,2,4,4,6,6-Hexamethylcyclotrisilazane ≥97.0% (by GC, titration analysis)
2,2,4,4,6,6-Hexamethylcyclotrisilazane

CAS Number: 1009-93-4
MDL Number: MFCD00039698
Molecular Formula: C6H21N3Si3
Molecular Weight: 219.51
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 188
Flash Point (°C): 61
Freezing point (°C): -10
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Dimethylsilazane Cyclic Trimer

Formula: C₆H₂₁N₃Si₃
MW: 219.51 g/mol
Boiling Pt: 188 °C
Density: 0.92 (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00039698
CAS Number: 1009-93-4
EINECS: 213-773-6
https://us.vwr.com/stibo/web/std.lang.all/09/31/28970931.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR