3-Heptylthiophene ≥94.0%

Supplier: TCI America

H0722-5G H0722-1G
TCH0722-005GEA 260 USD
TCH0722-005G TCH0722-001G
3-Heptylthiophene ≥94.0%
3-Heptylthiophene

CAS Number: 65016-61-7
MDL Number: MFCD00130131
Molecular Formula: C11H18S
Molecular Weight: 182.33
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Specific Gravity (20/20): 0.92

Formula: C₁₁H₁₈S
MW: 182.33 g/mol
Boiling Pt: 140 °C
Density: 0.92 (20 °C)
MDL Number: MFCD00130131
CAS Number: 65016-61-7
https://us.vwr.com/stibo/web/std.lang.all/07/61/28970761.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR