1,6-Heptadiene ≥99.0%

Supplier: TCI America

Synonyms: Hepta-1,6-diene

H0564-1ML H0564-5ML
TCH0564-005MLEA 365.29 USD
TCH0564-005ML TCH0564-001ML
1,6-Heptadiene ≥99.0%
1,6-Heptadiene

CAS Number: 3070-53-9
MDL Number: MFCD00008667
Molecular Formula: C7H12
Molecular Weight: 96.17
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Melting point (°C): -129
Flash Point (°C): -10
Specific Gravity (20/20): 0.71

Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 90 °C
Melting Pt: –129 °C
Density: 0.71 (20 °C)
Flash Pt: –10 °C
MDL Number: MFCD00008667
CAS Number: 3070-53-9
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR