1,2,5,6,9,10-Hexabromocyclododecane ≥97.0% (by titrimetric analysis)

H0544-25G
TCH0544-025GEA 22 USD
TCH0544-025G
1,2,5,6,9,10-Hexabromocyclododecane ≥97.0% (by titrimetric analysis)
1,2,5,6,9,10-Hexabromocyclododecane

CAS Number: 3194-55-6
MDL Number: MFCD00003716
Molecular Formula: C12H18Br6
Molecular Weight: 641.70
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 177

Formula: C₁₂H₁₈Br₆
MW: 641.7 g/mol
Boiling Pt: 462 °C (1013 hPa)
Melting Pt: 177 °C
CAS Number: 3194-55-6
EINECS: 221-695-9

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR