Hexadecylbenzene ≥99.0%

Supplier: TCI America

Synonyms: Cetylbenzene1-Phenylhexadecane

H0513-5G H0513-25G
TCH0513-5GEA 124.81 USD
TCH0513-5G TCH0513-025G
Hexadecylbenzene ≥99.0%
Hexadecylbenzene

CAS Number: 1459-09-2
MDL Number: MFCD00039871
Molecular Formula: C22H38
Molecular Weight: 302.55
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 172
Freezing point (°C): 25


Other Product Names / Synonyms: Cetylbenzene, 1-Phenylhexadecane

Formula: C₂₂H₃₈
MW: 302.54 g/mol
Boiling Pt: 172 °C
Melting Pt: 27 °C
Density: 0.8597 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039871
CAS Number: 1459-09-2
EINECS: 215-949-8
https://us.vwr.com/stibo/web/std.lang.all/05/22/28970522.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR